Skip to Content
Merck
All Photos(1)

Key Documents

240079

Sigma-Aldrich

Diphenylmethane

≥99%

Synonym(s):

Benzylbenzene, Methylenedibenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CH2
CAS Number:
Molecular Weight:
168.23
Beilstein:
1904982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.79 (vs air)

vapor pressure

<1 mmHg ( 77 °C)

Assay

≥99%

autoignition temp.

905 °F

refractive index

n20/D 1.577 (lit.)

bp

264 °C (lit.)

mp

22-24 °C (lit.)

solubility

alcohol: freely soluble(lit.)
benzene: freely soluble(lit.)
chloroform: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
hexane: freely soluble(lit.)
insoluble (liquid ammonia)(lit.)

density

1.006 g/mL at 25 °C (lit.)

SMILES string

C(c1ccccc1)c2ccccc2

InChI

1S/C13H12/c1-3-7-12(8-4-1)11-13-9-5-2-6-10-13/h1-10H,11H2

InChI key

CZZYITDELCSZES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenylmethane skeleton acted as a surrogate for a steroid skeleton.

Application

Diphenylmethane (Benzylbenzene) derivatives were used as estrogen receptor (ER) agonists and antagonists.

replaced by

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

266.0 °F

Flash Point(C)

130 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Maruyama et al.
Bioorganic & medicinal chemistry letters, 23(14), 4031-4036 (2013-06-19)
Our multi-template approach for drug discovery, focusing on protein targets with similar fold structures, has yielded lead compounds for various targets. We have also shown that a diphenylmethane skeleton can serve as a surrogate for a steroid skeleton. Here, on
Daisuke Kajita et al.
Bioorganic & medicinal chemistry, 22(7), 2244-2252 (2014-03-19)
Steroid sulfatase (STS) is a potential target for treatment of postmenopausal hormone-dependent breast cancer. Several steroidal STS inhibitors have been reported, but steroidal compounds are difficult to optimize and may interact with other targets. On the other hand, we have
Carsten Uerpmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(6), 1750-1756 (2005-01-26)
A new tetracationic triple-stranded supramolecular cylinder is prepared from a bis(pyridylimine) ligand containing a diphenylmethane and two ketimine groups in the spacer. The cylinder is longer and slightly wider than the corresponding cylinder containing just diphenylmethane spacers. Inter-strand CH...pi interactions
S H Safe et al.
Reproduction, fertility, and development, 13(4), 307-315 (2002-01-22)
It has been hypothesized that environmental contaminants that modulate endocrine signaling pathways may be causally linked to adverse health effects in humans. There has been particular concern regarding synthetic estrogens and their role in disrupting normal development of the male
Ben Nebgen et al.
The Journal of chemical physics, 137(8), 084112-084112 (2012-09-04)
The theory for modeling vibronic interactions in bichromophores was introduced in sixties by Witkowski and Moffitt [J. Chem. Phys. 33, 872 (1960)] and extended by Fulton and Gouterman [J. Chem. Phys. 35, 1059 (1961)]. The present work describes extension of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service