Skip to Content
Merck
All Photos(1)

Key Documents

238643

Sigma-Aldrich

IGEPAL® CO-520

average Mn 441

Synonym(s):

Polyoxyethylene (5) nonylphenylether, branched

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€58.30
500 G
€174.00

€58.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€58.30
500 G
€174.00

About This Item

Linear Formula:
(C2H4O)n · C15H24O, n~5
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€58.30


Check Cart for Availability

Request a Bulk Order

mol wt

average Mn 441

Quality Level

refractive index

n20/D 1.496 (lit.)

density

0.997 g/mL at 25 °C (lit.)

HLB

10

SMILES string

CCCCCCCCCc1ccc(O)cc1.OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C24H50O13.C15H24O/c25-1-3-27-5-7-29-9-11-31-13-15-33-17-19-35-21-23-37-24-22-36-20-18-34-16-14-32-12-10-30-8-6-28-4-2-26;1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h25-26H,1-24H2;10-13,16H,2-9H2,1H3

InChI key

SUJLMNLVFQRPGM-UHFFFAOYSA-N

General description

IGEPAL®CO-520 or poly (oxyethylene)nonylphenyl ether) is a non-ionic surfactant.

Application

Igepal aggregates have been used in fluorescence and light-scattering studies.[1]Used in the preparation of monodispersed colloidal silica cadmium suphide nanocomposites,[2] alumina nanoparticles,[3] ultra thin silica coated magnetic nanoparticles.[4]

Legal Information

IGEPAL is a registered trademark of Solvay

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

380.3 °F - closed cup

Flash Point(C)

193.5 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aggregation of non ionic surfactant Igepal in aqueous solution: fluorescence and light scattering studies
Ghosh SK, et al.
International Journal of Molecular Sciences, 4, 562-571 (2003)
Synthesis and characterization of alumina nanoparticles by igepal CO-520 stabilized reverse micelle and sol-gel processing
Chandradass J and Bae DS
Materials and Manufacturing Processes, 23(5), 494-498 (2008)
Preparation and properties of tailored morphology, monodisperse colloidal silica-cadmium sulfide nanocomposites.
Chang SY, et al.
Journal of the American Chemical Society, 116(15), 6739-6744 (1994)
Ming Zhang et al.
Nanotechnology, 19(8), 085601-085601 (2008-02-27)
A systematic study of the formation of silica-coated magnetite particles via a modified reverse microemulsion approach was investigated by using transmission electron microscopy (TEM), x-ray diffraction (XRD) and a superconducting quantum interference device (SQUID). The results show that the surfactant
Mirko Trilling et al.
Journal of virology, 83(8), 3684-3695 (2009-02-13)
Vaccinia virus (VACV) replicates in mouse and human fibroblasts with comparable kinetics and efficiency, yielding similar titers of infectious progeny. Here we demonstrate that gamma interferon (IFN-gamma) but not IFN-alpha or IFN-beta pretreatment of mouse fibroblasts prior to VACV infection

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service