Skip to Content
Merck
All Photos(1)

Key Documents

237701

Sigma-Aldrich

S-Alpine-Borane® solution

0.5 M in THF

Synonym(s):

B-isopinocampheyl-9-borabicyclo[3.3.1]nonane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31B
CAS Number:
Molecular Weight:
258.25
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

liquid

optical activity

[α]22/D +1.2°, neat

concentration

0.5 M in THF

density

0.897 g/mL at 25 °C

SMILES string

C[C@@H]1[C@H](C[C@H]2C[C@@H]1C2(C)C)B3[C@@H]4CCC[C@H]3CCC4

InChI

1S/C18H31B/c1-12-16-10-13(18(16,2)3)11-17(12)19-14-6-4-7-15(19)9-5-8-14/h12-17H,4-11H2,1-3H3/t12-,13+,14-,15+,16-,17-/m0/s1

InChI key

VCDGSBJCRYTLNU-NEXGVSGLSA-N

General description

S-Alpine-Borane® is a chiral reducing agent, synthesized from (-)-α-pinene via hydroboration.

Application

R- and S-Alpine-Boranes are used for asymmetric reduction of aldehydes and prochiral ketones.
S-Alpine-Borane® solution may be used to prepare:
  • 5(S)-methyl 5-hydroxy-8-(tetrahydropyran-2-yloxy)-6-octynoate, an intermediate for the stereocontrolled total synthesis of leukotriene B4.
  • (+)-(S)-4-hydroxy-6-phenyl-5-hexynyl 2,2,4,4-tetramethyl-1,3-oxazolidine-3-carboxylate
  • (R)-1-(tert-Butyldiphenylsilyloxy)pent-3-yn-2-ol, an intermediate for the preparation of (S)-4,5-dihydroxypentane-2,3-dione (DPD).

Legal Information

Alpine-Borane is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Pyr. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of the precursor of AI-2, the signalling molecule for inter-species quorum sensing.
Ascenso OS, et al.
Bioorganic & Medicinal Chemistry, 19(3), 1236-1241 (2011)
(-)-Sparteine-Mediated Stereoselective Intramolecular Carbolithiation of 4-Substituted 5-Hexynyl Carbamates. Synthesis of Enantiopure 1, 3-Difunctionalized Alkylidene Cyclopentanes.
Oestreich M, et al.
The Journal of Organic Chemistry, 64(23), 8616-8626 (1999)
Tetrahedron Asymmetry, 5, 1061-1061 (1994)
Total synthesis of leukotriene B4.
Kerdesky FAJ, et al.
The Journal of Organic Chemistry, 58(13), 3516-3520 (1993)
Synlett, 561-561 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service