237604
4-Hexen-1-ol, predominantly trans
97%
Synonym(s):
(4E)-4-Hexen-1-ol, (E)-4-Hexen-1-ol, trans-4-Hexen-1-ol, trans-4-Hexenol
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.439 (lit.)
bp
159-160 °C (lit.)
density
0.851 g/mL at 25 °C (lit.)
functional group
hydroxyl
SMILES string
C\C=C\CCCO
InChI
1S/C6H12O/c1-2-3-4-5-6-7/h2-3,7H,4-6H2,1H3/b3-2+
InChI key
VTIODUHBZHNXFP-NSCUHMNNSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
141.8 °F - closed cup
Flash Point(C)
61 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service