Skip to Content
Merck
All Photos(1)

Key Documents

235822

Sigma-Aldrich

3-Fluorobenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CN
CAS Number:
Molecular Weight:
121.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5043 (lit.)

bp

182-183 °C/753 mmHg (lit.)

mp

−16 °C (lit.)

density

1.133 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

Fc1cccc(c1)C#N

InChI

1S/C7H4FN/c8-7-3-1-2-6(4-7)5-9/h1-4H

InChI key

JZTPKAROPNTQQV-UHFFFAOYSA-N

General description

Microwave spectrum of 3-fluorobenzonitrile has been studied[1]. Amination of 3-fluorobenzonitrile with morpholine in DMSO at 100°C has been investigated[2].

Application

3-Fluorobenzonitrile has been used in the preparation of bis(3-cyanophenoxy)phenylenes[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved yields of meta-amination and symmetrical and unsymmetrical diamination of benzenes.
Brown GR, et al.
Tetrahedron Letters, 42(23), 3917-3919 (2001)
Microwave spectral study of 3-fluorobenzonitrile.
Dutta A, et al.
Journal of Molecular Spectroscopy, 118(1), 232-236 (1986)
Xiaotong Cheng et al.
The Journal of clinical investigation, 130(5), 2237-2251 (2020-01-31)
Hypoxia-inducible factor (HIF) is strikingly upregulated in many types of cancer, and there is great interest in applying inhibitors of HIF as anticancer therapeutics. The most advanced of these are small molecules that target the HIF-2 isoform through binding the
Synthesis of bis (ether nitrile) s and bis (ether acid) s from simplearomatic diols bymeta-fluoro displacement from3-fluorobenzonitrile.
Eastmond GC and Paprotny J.
Journal of Materials Chemistry, 7(8), 1321-1326 (1997)
Oriol Planas et al.
Science (New York, N.Y.), 367(6475), 313-317 (2020-01-18)
Bismuth catalysis has traditionally relied on the Lewis acidic properties of the element in a fixed oxidation state. In this paper, we report a series of bismuth complexes that can undergo oxidative addition, reductive elimination, and transmetallation in a manner

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service