Skip to Content
Merck
All Photos(1)

Key Documents

235032

Sigma-Aldrich

Bis(trimethylsilyl) malonate

98%

Synonym(s):

Malonic acid bis(trimethylsilyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOCOCH2COOSi(CH3)3
CAS Number:
Molecular Weight:
248.42
Beilstein:
2212115
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.416 (lit.)

bp

63-66 °C/1 mmHg (lit.)

density

0.974 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C

InChI

1S/C9H20O4Si2/c1-14(2,3)12-8(10)7-9(11)13-15(4,5)6/h7H2,1-6H3

InChI key

ATCKJLDGNXGLAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(trimethylsilyl) malonate is precursor for metal-organic chemical vapor deposition (MOCVD) of HfO2 and ZrO2 thin films[1]. It undergoes acylation reaction with acid chlorides and acyl carbonates in the presence of triethylamine and magnesium or lithium salts to give β-keto acids or methyl ketones[2].

Application

Bis(trimethylsilyl) malonate was used as cyclocondensation agent in cyclocondensation reactions[3]. It was also used in the preparation of β-keto acids and methyl ketones[4].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malonates in cyclocondensation reactions.
Stadlbauer W, et al.
Molecules (Basel), 6(4), 338-352 (2001)
Synthesis of ?-Keto Acids and Methyl Ketones Using Bis (trimethylsilyl) Malonate and Triethylamine in the Presence of Lithium or Magnesium Halides.
Rathke MW and Nowak MA.
Synthetic Communications, 15(12), 1039-1049 (1985)
Ramasamy Pothiraja et al.
Dalton transactions (Cambridge, England : 2003), (4)(4), 654-663 (2009-04-22)
New Hf and Zr malonate complexes have been synthesized by the reaction of metal amides with different malonate ligands (L = dimethyl malonate (Hdmml), diethyl malonate (Hdeml), di-tert-butyl malonate (Hdbml) and bis(trimethylsilyl) malonate (Hbsml)). Homoleptic eight-coordinated monomeric compounds of the
Synthetic Communications, 15, 1039-1039 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service