Skip to Content
Merck
All Photos(1)

Key Documents

233234

Sigma-Aldrich

3-Chloro-4-fluoronitrobenzene

98%

Synonym(s):

2-Chloro-1-fluoro-4-nitrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€160.00

€160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€160.00

About This Item

Linear Formula:
ClC6H3(F)NO2
CAS Number:
Molecular Weight:
175.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€160.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

40-42 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)c(Cl)c1

InChI

1S/C6H3ClFNO2/c7-5-3-4(9(10)11)1-2-6(5)8/h1-3H

InChI key

DPHCXXYPSYMICK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-1-fluoro-4-nitrobenzene undergoes reduction in the presence of Mo(CO)6 and 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU) under microwave irradiation to yield a mixture of 3-chloro-4-fluoroaniline and 3-chloro-4-ethoxyaniline[1].

Application

2-Chloro-1-fluoro-4-nitrobenzene has been used in synthesis of:
  • benzoimidolones on Ameba resin[2]
  • methyl 6,7,8,9-tetrahydropyrido[1,2-a]indol-10-ylacetate[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical and efficient synthesis of N-fused tricyclic indoles.
Koay N, et al.
Tetrahedron Letters, 52(1), 122-124 (2011)
Microwave mediated reduction of heterocycle and fluorine containing nitroaromatics with Mo (CO)6 and DBU.
Spencer J, et al.
Tetrahedron, 64(44), 10195-10200 (2008)
Microwave-assisted traceless synthesis of benzimidazolones.
Xu X-J and Zong Y-X.
Tetrahedron Letters, 48(1), 129-132 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service