232726
8-Methoxypsoralen
99%
Synonym(s):
8-MOP, 9-Methoxyfuro[3,2-g][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin
About This Item
Recommended Products
Assay
99%
mp
148-150 °C (lit.)
SMILES string
COc1c2OC(=O)C=Cc2cc3ccoc13
InChI
1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3
InChI key
QXKHYNVANLEOEG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service