Skip to Content
Merck
All Photos(1)

Key Documents

232343

Sigma-Aldrich

4-Butoxyaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC6H4NH2
CAS Number:
Molecular Weight:
165.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

refractive index

n20/D 1.538 (lit.)

bp

148-149 °C/13 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

SMILES string

CCCCOc1ccc(N)cc1

InChI

1S/C10H15NO/c1-2-3-8-12-10-6-4-9(11)5-7-10/h4-7H,2-3,8,11H2,1H3

InChI key

UBRIHZOFEJHMIT-UHFFFAOYSA-N

General description

4-Butoxyaniline undergoes low-pressure hydrogenation in the presence of rhodium on alumina to yield 4-butoxycyclohexylamine[1].

Application

4-Butoxyaniline was used in the preparation of:
  • bis-4-butoxyphenyl-4,4′-(2,2′-bipyridyl)carbamide[2]
  • 1-(4-butoxyphenyl)-3-(6-methyl-3-oxo-2-phenyl-2,3-dihydropyridazin-4-yl)urea[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkali metal cation cooperative anion recognition by heteroditopic bis (calix [4] arene) rhenium (I) bipyridyl and ferrocene receptor molecules.
Cooper JB, et al.
J. Chem. Soc., Dalton Trans., 16, 2721-2728 (2000)
Low-Pressure Hydrogenation of Alkoxyanilines with Noble Metal Catalysts.
Freifelder M, et al.
The Journal of Organic Chemistry, 30(7), 2485-2486 (1965)
Agostino Cilibrizzi et al.
Journal of medicinal chemistry, 52(16), 5044-5057 (2009-07-31)
Following a ligand-based drug design approach, a potent mixed formyl peptide receptor 1 (FPR1) and formyl peptide receptor-like 1 (FPRL1) agonist (14a) and a potent and specific FPRL1 agonist (14x) were identified. These compounds belong to a large series of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service