Skip to Content
Merck
All Photos(2)

Documents

230103

Sigma-Aldrich

2,4-Diaminophenol dihydrochloride

98%

Synonym(s):

‘Amidol’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H3OH·2HCl
CAS Number:
Molecular Weight:
197.06
Beilstein:
3913728
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

222 °C (dec.) (lit.)

SMILES string

Cl.Cl.Nc1ccc(O)c(N)c1

InChI

1S/C6H8N2O.2ClH/c7-4-1-2-6(9)5(8)3-4;;/h1-3,9H,7-8H2;2*1H

InChI key

KQEIJFWAXDQUPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Diaminophenol dihydrochloride (Amidol) was used in the dye manufacturing industries and as a color accelerator in photographic developers.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

National Toxicology Program technical report series, 401, 1-232 (1992-05-01)
2,4-Diaminophenol dihydrochloride is used in the manufacture of dyes and as a color accelerator in photographic developers. Toxicology and carcinogenesis studies were conducted by administering 2,4-diaminophenol dihydrochloride (greater than 97% pure) in corn oil by gavage to F344/N rats and
M Kh Saiatov et al.
Voprosy virusologii, 30(1), 39-43 (1985-01-01)
The method of antibody sensitization of sheep erythrocyte using amidole was employed to prepare diagnosticums for influenza A/USSR/090/77 (H1N1), A/Bangkok/1/79 (H3N2), and B/Hong Kong/75 viruses. Comparison of the potency of diagnosticums obtained on the basis of hyperimmune influenza sera and
Caroline Fribourg et al.
Biochemistry, 45(35), 10777-10785 (2006-08-31)
The light chain of activated factor IX (FIXa) is involved in a number of functional properties, including FIXa enzymatic activity. This suggests the existence of a functional link between the FIXa light chain and the catalytic domain. The FIXa structure
[Preparation of antibody erythrocyte for indicating arbovirus antigens].
T V Kiriushchenko et al.
Voprosy virusologii, (1)(1), 98-102 (1982-01-01)
V A Shamardin
Journal of hygiene, epidemiology, microbiology, and immunology, 27(3), 265-270 (1983-01-01)
Selection of the method of loading erythrocytes with preparations of immunoglobulins (IG) should be determined by the properties of the sensitins to be used. For the production of efficient antibody diagnostics it is advisable to use amidol while in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service