Skip to Content
Merck
All Photos(2)

Key Documents

229784

Sigma-Aldrich

Manganese(II) sulfate hydrate

≥99.99% trace metals basis

Synonym(s):

Manganous(II) sulfate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€199.00
100 G
€554.00

€199.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€199.00
100 G
€554.00

About This Item

Linear Formula:
MnSO4·xH2O
CAS Number:
Molecular Weight:
151.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

€199.00


Check Cart for Availability

Request a Bulk Order

Agency

suitable for SM 5210

Quality Level

Assay

≥99.99% trace metals basis

form

solid

impurities

≤100.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

storage temp.

2-8°C

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II) sulfate hydrate is a water-soluble solid with diverse industrial applications. It is utilized in the production of fertilizers, ceramics, and other manganese compounds. Furthermore, it serves as an electrolyte additive in lithium-ion and alkaline batteries to improve their capacity and stability. Additionally, it is employed in the manufacture of thin-film solar cells, contributing to their efficiency and stability.

Application

Manganese(II) sulfate hydrate can be in research and development:
  • As an electrolyte additive in rechargeable aqueous zinc/sodium manganese oxides batteries. It helps to stabilize the manganese oxide cathode and improve the overall electrochemical stability of the battery      
  • In the synthesis of electrodes for supercapacitors due to its high theoretical capacitance, low cost, and environmental friendliness.
  • As a precursor material to synthesize various forms of manganese oxide (MnOx) for use as electrode materials in supercapacitors.
  • As a precursor materials along with other transition metal sulfates for the synthesis of cathode materials such as NMC for Lithium-ion batteries.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jimmy E Becerra et al.
Applied and environmental microbiology, 81(11), 3880-3888 (2015-03-31)
L-Fucose is a sugar present in human secretions as part of human milk oligosaccharides, mucins, and other glycoconjugates in the intestinal epithelium. The genome of the probiotic Lactobacillus rhamnosus GG (LGG) carries a gene cluster encoding a putative L-fucose permease
Olivier Biner et al.
PloS one, 10(12), e0143963-e0143963 (2015-12-03)
Hyaluronidases are important venom components acting as spreading factor of toxic compounds. In several studies this spreading effect was tested on vertebrate tissue. However, data about the spreading activity on invertebrates, the main prey organisms of spiders, are lacking. Here

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service