Select a Size
About This Item
acetone: slightly soluble(lit.)
diethyl ether: slightly soluble(lit.)
ethanol: slightly soluble(lit.)
Recommended Products
grade
for analytical purposes
Quality Level
Assay
99%
form
solid
reaction suitability
reagent type: catalyst
core: cadmium
bp
787 °C (lit.)
mp
388 °C (lit.)
solubility
H2O: very soluble(lit.)
acetone: slightly soluble(lit.)
diethyl ether: slightly soluble(lit.)
ethanol: slightly soluble(lit.)
density
5.67 g/mL at 25 °C (lit.)
SMILES string
I[Cd]I
InChI
1S/Cd.2HI/h;2*1H/q+2;;/p-2
InChI key
OKIIEJOIXGHUKX-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service