226955
2-(Triphenylphosphoranylidene)succinic anhydride
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H17O3P
CAS Number:
Molecular Weight:
360.34
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Assay
97%
reaction suitability
reaction type: C-C Bond Formation
mp
167 °C (dec.) (lit.)
SMILES string
O=C1CC(C(=O)O1)=P(c2ccccc2)(c3ccccc3)c4ccccc4
InChI
1S/C22H17O3P/c23-21-16-20(22(24)25-21)26(17-10-4-1-5-11-17,18-12-6-2-7-13-18)19-14-8-3-9-15-19/h1-15H,16H2
InChI key
CXYNGWAHZKOMJQ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service