Skip to Content
Merck
All Photos(3)

Key Documents

225134

Sigma-Aldrich

Ethyl (2-fluorobenzoyl)acetate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
210.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

liquid

refractive index

n20/D 1.516 (lit.)

density

1.164 g/mL at 25 °C (lit.)

functional group

ester
fluoro
ketone

SMILES string

CCOC(=O)CC(=O)c1ccccc1F

InChI

1S/C11H11FO3/c1-2-15-11(14)7-10(13)8-5-3-4-6-9(8)12/h3-6H,2,7H2,1H3

InChI key

YMUNUVJSNWUWDA-UHFFFAOYSA-N

Application

Ethyl (2-fluorobenzoyl)acetate may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service