Skip to Content
Merck
All Photos(1)

Documents

223964

Sigma-Aldrich

Bis(tetrabutylammonium) dichromate

99%

Synonym(s):

TBADC, Tetrabutylammonium dichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3CH2CH2CH2)4N]2Cr2O7
CAS Number:
Molecular Weight:
700.92
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

reaction suitability

reagent type: oxidant

mp

139-143 °C (lit.)

SMILES string

[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/2C16H36N.2Cr.7O/c2*1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;;;;;;/h2*5-16H2,1-4H3;;;;;;;;;/q2*+1;;;;;;;;2*-1

InChI key

MMKQSIQNDMIVIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(tetrabutylammonium) dichromate (TBADC) is a mild oxidizing agent which can be used as a reagent for:
  • Preparation of nitrophenol derivatives and quinones under aprotic reaction conditions.
  • Conversion of oximes to carbonyl compounds under microwave irradiation.
  • Catalyst precursor for gas phase dehydration of acetic acid to ketene.

Reagent for:
  • Preparation of nitrophenol derivatives and quinones under aprotic reaction conditions
  • Conversion of oximes to carbonyl compounds under microwave irradiation

  • Catalyst precursor for catalytic gas phase dehydration of acetic acid to ketene
  • Crosslinking agent

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitration of phenolic compounds and oxidation of hydroquinones using tetrabutylammonium chromate and dichromate under aprotic conditions.
Pourali AR and Goli A
Journal of Chemical Sciences (Bangalore), 123(1), 63-67 (2011)
Regeneration of carbonyl compounds from oximes using BTBAD under microwave irradiation.
Murugan R and Reddy BSR
Chemistry Letters (Jpn), 33(8), 1038-1039 (2004)
Bis-tetrabutylammonium Dichromate as a Neutral and Selective Oxidant of Hydroxy Compounds.
Santaniello E and Ferraboschi P
Synthetic Communications, 10(1), 75-81 (1980)
Catalytic gas phase dehydration of acetic acid to ketene.
Waters T, et al.
International Journal of Mass Spectrometry, 228(2-3), 599-611 (2003)
Bis (tetrabutylammonium) Dichromate.
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service