222852
D-Ornithine monohydrochloride
98%
Synonym(s):
(R)-(−)-2,5-Diaminopentanoic acid hydrochloride, (R)-2,5-Diaminopentanoic acid monohydrochloride, D-2,5-Diaminopentanoic acid monohydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
optical activity
[α]22/D −21.8°, c = 2 in 5 M HCl
optical purity
ee: 96% (GLC)
mp
239 °C (dec.) (lit.)
SMILES string
Cl.NCCC[C@@H](N)C(O)=O
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service