Skip to Content
Merck
All Photos(1)

Key Documents

222704

Sigma-Aldrich

2-Fluoro-4-nitrotoluene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€96.00

€96.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€96.00

About This Item

Linear Formula:
CH3C6H3(NO2)F
CAS Number:
Molecular Weight:
155.13
Beilstein:
2250156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€96.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

65-68 °C/2 mmHg (lit.)

mp

31-35 °C (lit.)

functional group

fluoro
nitro

SMILES string

Cc1ccc(cc1F)[N+]([O-])=O

InChI

1S/C7H6FNO2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,1H3

InChI key

WIQISTBTOQNVCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Fluoro-4-nitrotoluene was used in the synthesis of 2-fluoro-4-nitrobenzoic acid[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

359. Preparation of some fluoronitro-and aminofluoro-benzoic acids, and of fluoronitro-benzenes,-toluenes, and-xylenes.
Journal of the Chemical Society, 1925-1927 (1963)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service