Recommended Products
composition
Al-Ni, 50:50 wt. %
reaction suitability
core: nickel
reagent type: catalyst
manufacturer/tradename
W.R. Grace and Co. Raney®
density
3.46 g/mL at 25 °C (lit.)
SMILES string
[Al].[Ni]
InChI
1S/Al.Ni
InChI key
NPXOKRUENSOPAO-UHFFFAOYSA-N
Related Categories
Application
- Reduction reactions
- Reductive dehalogenation and ring saturation reactions
Used for:
- Preparation of aluminum oxy-hydrate supported NiAl skeleton catalyst for hydrogenation reactions
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Flam. Sol. 1 - Skin Sens. 1 - Water-react 2
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service