Skip to Content
Merck
All Photos(1)

Key Documents

220698

Sigma-Aldrich

L-Valine ethyl ester hydrochloride

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€60.90

€60.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€60.90

About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2C2H5 · HCl
CAS Number:
Molecular Weight:
181.66
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€60.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

optical activity

[α]20/D +6.7°, c = 2 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

102-105 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].CCOC(=O)[C@@H](N)C(C)C

InChI

1S/C7H15NO2.ClH/c1-4-10-7(9)6(8)5(2)3;/h5-6H,4,8H2,1-3H3;1H/t6-;/m0./s1

InChI key

PQGVTLQEKCJXKF-RGMNGODLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon J Dunne et al.
Drug testing and analysis, 9(5), 734-743 (2016-07-13)
Herbal smoking blends containing synthetic cannabinoids have become popular alternatives to marijuana. These products were previously sold in pre-packaged foil bags, but nowadays seizures usually contain synthetic cannabinoid powders together with unprepared plant materials. A question often raised by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service