Skip to Content
Merck
All Photos(1)

Key Documents

220108

Sigma-Aldrich

Isovanillic acid

97%

Synonym(s):

3-Hydroxy-4-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
168.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

250-253 °C (lit.)

functional group

carboxylic acid

SMILES string

COc1ccc(cc1O)C(O)=O

InChI

1S/C8H8O4/c1-12-7-3-2-5(8(10)11)4-6(7)9/h2-4,9H,1H3,(H,10,11)

InChI key

LBKFGYZQBSGRHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rate of oxidation of isovanillic acid by washed cells of Pseudomonas putida has been reported[1].

Application

Isovanillic acid has been used in the synthesis of opiate alkaloid precursor, dihydrothebainone[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Jeffers et al.
Microbiology (Reading, England), 143 ( Pt 6), 1975-1981 (1997-06-01)
A methyltransferase enzyme catalysing the 3-O-methylation of isovanillic acid (3-hydroxy-4-methoxybenzoic acid) by S-adenosylmethionine (SAM) was identified in Phanerochaete chrysosporium and purified. Gel filtration indicated an M(r) of 71,000 and SDS-PAGE showed that the enzyme was composed of two subunits of
Preparation of oxygenated phenylacetic acids.
Weller DD, et al.
The Journal of Organic Chemistry, 49(11), 2061-2063 (1984)
Widiastuti Setyaningsih et al.
Food chemistry, 288, 221-227 (2019-03-25)
Fifteen phenolic compounds were determined in rice grains by ultrasound assisted extraction and ultra-performance liquid chromatography with photodiode array detection. Primarily, an UAE method has been developed and validated for the extraction of phenolics from rice grains. For the optimization
Motohiro Nishimura et al.
Journal of bioscience and bioengineering, 102(2), 124-127 (2006-10-10)
Strain NL15-2K was isolated from soil by screening for bacteria capable of catabolizing lignin-related aromatic acids. This isolate was identified as a Streptomyces sp. on the basis of morphology and an analysis of its 16S rRNA gene sequence. NL15-2K utilized
I Reenilä et al.
Journal of chromatography. B, Biomedical applications, 663(1), 137-142 (1995-01-06)
We applied coulometric detection (three electrochemical electrodes in series) to quantitate vanillic acid and isovanillic acid using reversed-phase HPLC. The formation of these reaction products from dihydroxybenzoic acid was used as a precise and reproducible measure of catechol-O-methyltransferase (COMT) activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service