Skip to Content
Merck
All Photos(2)

Documents

220078

Sigma-Aldrich

Isethionic acid sodium salt

98%

Synonym(s):

2-Hydroxyethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2SO3Na
CAS Number:
Molecular Weight:
148.11
Beilstein:
3633992
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

191-194 °C (lit.)

SMILES string

[Na+].OCCS([O-])(=O)=O

InChI

1S/C2H6O4S.Na/c3-1-2-7(4,5)6;/h3H,1-2H2,(H,4,5,6);/q;+1/p-1

InChI key

LADXKQRVAFSPTR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Zhong et al.
PloS one, 6(2), e16970-e16970 (2011-03-09)
Serotonin exerts a powerful influence on neuronal excitability. In this study, we investigated the effects of serotonin on different neuronal populations in prefrontal cortex (PFC), a major area controlling emotion and cognition. Using whole-cell recordings in PFC slices, we found
Judy A Libra et al.
Chemosphere, 56(2), 167-180 (2004-05-04)
A two stage anaerobic/aerobic bacterial process was used to decolorize and partially mineralize a reactive vinyl sulfone diazo dye C.I. Reactive Black 5 (RB5) in a synthetic wastewater. Since the anchor group of reactive dyes reacts during the dyeing process
Lei Zhang et al.
Biosensors & bioelectronics, 24(4), 690-695 (2008-08-19)
A composite film of polyaniline (PAN) nano-networks/p-aminobenzene sulfonic acid (ABSA) modified glassy carbon electrode (GCE) has been fabricated via an electrochemical oxidation procedure and applied to the electro-catalytic oxidation of uric acid (UA) and ascorbic acid (AA). The ABSA monolayer
S Kearns et al.
Advances in experimental medicine and biology, 483, 563-570 (2002-01-15)
Taurine has been shown to be an effective scavenger of hypochlorous acid (HOCl). The role of HOCl is well established in tissue damage associated with reperfusion injury mediated by neutrophils. The role of HOCl in CNS injury and inflammatory reactions
Wei Tang et al.
Drug metabolism and disposition: the biological fate of chemicals, 30(7), 778-787 (2002-06-18)
(R)-N-[4-[2-[[2-Hydroxy-2-(pyridin-3-yl)ethyl]amino]ethyl]phenyl]- 4-[4-(4-trifluoro-methylphenyl)thiazol-2-yl]benzenesulfonamide (1) is a potent and selective agonist of the human beta3-adrenergic receptor. We report herein the data from studies of the metabolism and excretion of 1 in rats. Five metabolites were identified in the bile of male Sprague-Dawley

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service