Skip to Content
Merck
All Photos(1)

Key Documents

219193

Sigma-Aldrich

3-Thiopheneacetonitrile

96%

Synonym(s):

3-Thienylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NS
CAS Number:
Molecular Weight:
123.18
Beilstein:
107774
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.545 (lit.)

bp

124-125 °C/16 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

N#CCc1ccsc1

InChI

1S/C6H5NS/c7-3-1-6-2-4-8-5-6/h2,4-5H,1H2

InChI key

GWZCLMWEJWPFFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Thiopheneacetonitrile was used in the synthesis of:
  • (E)-3-phenyl-2-(3-thienyl) acrylonitrile
  • (Z)-3-(4-methylphenyl)-2-(3-thienyl) acrylonitrile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

246.2 °F - closed cup

Flash Point(C)

119 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(Z)-3-(4-Methylphenyl)-2-(3-thienyl) acrylonitrile.
Cobo D, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(!1), o5179-o5180 (2011)
Electrochemical characterization of poly 3-thiopheneacetonitrile: Technique, solvent and relaxation effects.
El-Abdallah GM.
Journal of Saudi Chemical Society (2011)
Debora Cobo et al.
Acta crystallographica. Section C, Crystal structure communications, 62(Pt 9), o550-o553 (2006-09-07)
The structure of (E)-2-(2-thienyl)-3-(3,4,5-trimethoxyphenyl)acrylonitrile, C16H15NO3S, contains no direction-specific intermolecular interactions. The molecules of (E)-3-(4-bromophenyl)-2-(2-thienyl)acrylonitrile, C13H8BrNS, exhibit orientational disorder of the thienyl fragment, and the molecules are linked into simple C(5) chains by a single C-H...N hydrogen bond. In (E)-3-phenyl-2-(3-thienyl)acrylonitrile, C13H9NS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service