Skip to Content
Merck
All Photos(2)

Key Documents

218758

Sigma-Aldrich

1-Chloro-3-nitrobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
1907947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

autoignition temp.

500 °F

bp

236 °C (lit.)

mp

43-47 °C (lit.)

solubility

alcohol: freely soluble (hot)(lit.)
alcohol: very slightly soluble(lit.)
carbon disulfide: freely soluble(lit.)
chloroform: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
glacial acetic acid: freely soluble(lit.)
water: insoluble(lit.)

density

1.534 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(Cl)c1

InChI

1S/C6H4ClNO2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H

InChI key

KMAQZIILEGKYQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Chloro-3-nitrobenzene was used in the preparation of aromatic azo compounds using gold (Au) nanoparticles supported on TiO2 as a catalyst[1]. It was also used as surrogate standard to evaluate the removal of 2,4-dinitrotoluene from contaminated soils using enhanced electrokinetic remediation[2].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

255.2 °F

Flash Point(C)

124 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclodextrin-enhanced extraction and removal of 2, 4-dinitrotoluene from contaminated soils.
Khodadoust AP, et al.
Environmental Engineering Science, 25(4), 615-626 (2008)
[Hygienic study of the stability of methylnitrophos and pentachloronitrobenzene used in agriculture].
Sh T Atabaev et al.
Gigiena i sanitariia, (2)(2), 74-75 (1983-02-01)
Violeta Simic-Milosevic et al.
Angewandte Chemie (International ed. in English), 48(22), 4061-4064 (2009-04-01)
Chirality chameleon: Inelastic electron tunneling manipulation can be used to change a single chloronitrobenzene (ClNB) molecule, randomly adsorbed on Au(111), into its desired enantiomeric form (r or l, see STM images and ball-and-stick representation) and to vary its rotational orientation.
Abdessamad Grirrane et al.
Nature protocols, 5(3), 429-438 (2010-03-06)
This protocol describes the aerobic oxidation of aromatic anilines to aromatic azo compounds using gold (Au) nanoparticles supported on TiO(2) as a catalyst. Yields above 98% are achieved under a few bars of oxygen pressure. It should be noted that
V Simic-Milosevic et al.
Physical review letters, 98(11), 116102-116102 (2007-05-16)
A scanning tunneling microscope operating at 5 K is used to induce the isomerization of single chloronitrobenzene molecules on Cu(111) and verify the reaction. The threshold voltage of (227+/-7) mV for this reaction is explained based on electron-induced vibrational heating.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service