Skip to Content
Merck
All Photos(1)

Key Documents

21850

Sigma-Aldrich

Carbonate on polymer support

extent of labeling: ~3.5 mmol/g NaCO3- loading

Synonym(s):

Amberlite IRA 900 NaCO3--form

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352128
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

~3.5 mmol/g NaCO3- loading

impurities

~20% water

Other Notes

Polymeric reagent for the conversion of alkyl halides to alcohols in an aprotic solvent ; Synthesis of oxazolidinones and aziridines

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Cardillo et al.
Journal of the Chemical Society. Chemical Communications, 1309-1309 (1982)
Margaret L Eng et al.
Ecotoxicology and environmental safety, 179, 104-110 (2019-04-27)
Tetrabromobisphenol A bis(2,3-dibromopropyl) ether (TBBPA-BDBPE) is an additive flame retardant used in polyolefins and polymers. It has been detected in biota, including in avian eggs, yet little is known of its effects. We assessed the pattern of TBBPA-BDBPE concentrations in
A. Bongini et al.
Journal of the Chemical Society. Perkin Transactions 1, 1339-1339 (1986)
Margaret L Eng et al.
Ecotoxicology and environmental safety, 179, 151-159 (2019-04-30)
Tetrabromobisphenol A bis(2,3-dibromopropyl ether) (TBBPA-BDBPE) and 1,2-bis(2,4,6-tribromophenoxy)ethane (BTPBE) are both brominated flame retardants (BFRs) that have been detected in birds; however, their potential biological effects are largely unknown. We assessed the effects of embryonic exposure to TBBPA-BDBPE and BTBPE in
Elena Chiurchiù et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-19)
Highly functionalized furans are the key scaffolds of many pharmaceuticals and bioactive natural products. Herein, we disclose a new fruitful synthesis of polyfunctionalized furans starting from β-nitroenones and α-functionalized ketones. The protocol involves two steps promoted by solid supported species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service