Skip to Content
Merck
All Photos(3)

Documents

218367

Sigma-Aldrich

(S)-(−)-Limonene

96%

Synonym(s):

(−)-p-Mentha-1,8-diene, (−)-Carvene, (S)-4-Isopropenyl-1-methyl cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2323991
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.7 (vs air)

vapor pressure

<3 mmHg ( 14.4 °C)
1 mmHg ( 20 °C)

Assay

96%

form

liquid

optical activity

[α]19/D −94°, c = 10 in ethanol

expl. lim.

6.1 %

refractive index

n20/D 1.471 (lit.)

bp

175-177 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CC(=C)[C@H]1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m1/s1

InChI key

XMGQYMWWDOXHJM-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

(S)-(-)-Limonene, a monoterpenoid compound, is mostly used in perfumery and in flavoring. On irradiation, it undergoes radiolysis, which leads to a reduction in the optical purity. (S)-(-)-Limonene also shows potent antimicrobial activity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gamma-radiolysis of chiral molecules: R (+)-limonene, S (-)-limonene and R (-)-a-phellandrene.
Cataldo F, et al.
J. Radioanal. Nucl. Chem., 262(2), 423-428 (2004)
Antimicrobial activity of limonene enantiomers and 1, 8-cineole alone and in combination.
Van Vuuren SF and Viljoen AM.
Flavour and Fragrance Journal, 22(6), 540-544 (2007)
Pamella Macedo de Souza et al.
Molecules (Basel, Switzerland), 22(4) (2017-04-20)
Strategies for the control of sulfate-reducing bacteria (SRB) in the oil industry involve the use of high concentrations of biocides, but these may induce bacterial resistance and/or be harmful to public health and the environment. Essential oils (EO) produced by
Malaria parasites produce volatile mosquito attractants.
Kelly M, et al.
mBio, 6(2), e00235-e00215 (2015)
P A Cornwell et al.
The Journal of pharmacy and pharmacology, 46(12), 938-950 (1994-12-01)
Wide-angle X-ray-diffraction experiments were used to investigate the molecular organization of barrier components of human stratum corneum. Diffraction lines related to the side-by-side lipid packing arrangements in the intercellular bilayers were identified as were patterns arising from secondary protein structures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service