Skip to Content
Merck
All Photos(1)

Key Documents

216690

Sigma-Aldrich

2,2,2-Triphenylacetophenone

98%

Synonym(s):

Benzopinacolone, Phenyl trityl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€37.00

€37.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€37.00

About This Item

Linear Formula:
(C6H5)3CCOC6H5
CAS Number:
Molecular Weight:
348.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€37.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

182-184 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(c1ccccc1)C(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C26H20O/c27-25(21-13-5-1-6-14-21)26(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

CFBBKHROQRFCNZ-UHFFFAOYSA-N

General description

Asymmetric reduction of 2,2,2-triphenylacetophenone using with potassium 9-O-(1,2: 5, 6-di-O-isopropylidene-α-D-glucofuranosyl)-9-boratabicyclo [3.3.1] nonane (chiral reducing agent) has been reported[1]. Dehydrative cyclization of 2,2,2-triphenylacetophenone has been reported[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral synthesis via organoboranes. 15. Selective reductions. 42. Asymmetric reduction of representative prochiral ketones with potassium 9-O-(1, 2: 5, 6-di-O-isopropylidene-. alpha.-D-glucofuranosyl)-9-boratabicyclo [3.3. 1] nonane.
Brown HC, etal.
The Journal of Organic Chemistry, 53(6), 1231-1238 (1988)
Douglas A Klumpp et al.
Applied catalysis. A, General, 336(1-2), 128-132 (2008-03-01)
The hydroxyalkylation reactions of aceanthrenequinone (6) and acenapthenequinone (7) with a series of arenes have been studied. In reactions with the Brønsted superacid CF(3)SO(3)H (triflic acid), the condensation products are formed in good yields (58-99%, 10 examples) with high regioselectivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service