Skip to Content
Merck
All Photos(3)

Key Documents

214353

Sigma-Aldrich

Palladium on calcium carbonate

greener alternative

extent of labeling: 5 wt. % loading, unreduced

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€95.30

€95.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€95.30

About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22

€95.30


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

autoignition temp.

>1120 °F

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

greener alternative category

SMILES string

[Ca++].[Pd++].[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/2CH2O3.Ca.Pd/c2*2-1(3)4;;/h2*(H2,2,3,4);;/q;;2*+2/p-4

InChI key

XGLHAVVVUOEGIO-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Catalyst for:
  • Aqueous Heck coupling and hydroarylation
  • Reduction reactions
  • Hydrogenation reactions
Palladium on calcium carbonate (Pd/CaCO3) can be used as a green catalyst for a variety of applications such as:
  • formation of an efficient catalytic system for Heck coupling reaction and hydroarylation reaction[1]
  • synthesis of Lindlar catalyst for selective hydrogenation of alkynes[2]
  • a catalytic reservoir in Miyaura-Suzuki cross-coupling reactions[3]
  • Stille cross-coupling reactions[4]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyaura-Suzuki cross-coupling reactions: the role of Pd/CaCO3 as catalyst reservoir
Oliveira BL and Antunes OAC
Letters in Organic Chemistry, 4(1), 13-15 (2007)
Palladium on Calcium Carbonate Combined to 2-Hydroxypropyl-alpha/beta-cyclodextrins: A Selective Catalytic System for Aqueous Heck Coupling and Hydroarylation
Senra JD, et al.
Advanced Synthesis & Catalysis, 350(16), 2551-2558 (2008)
Ligand-free Stille cross-coupling reaction using Pd/CaCO3 as catalyst reservoir
Coelho AV, et al.
Tetrahedron Letters, 48(43), 7671-7674 (2007)
Palladium on Calcium Carbonate Combined to 2-Hydroxypropyl-alpha/beta-cyclodextrins: A Selective Catalytic System for Aqueous Heck Coupling and Hydroarylation
Senra JD, et al.
advanced synthesis and catalysis, 350(16), 2551-2558 (2008)
Chun Wong Aaron Chan et al.
Nature communications, 5, 5787-5787 (2014-12-20)
Lindlar catalysts comprising of palladium/calcium carbonate modified with lead acetate and quinoline are widely employed industrially for the partial hydrogenation of alkynes. However, their use is restricted, particularly for food, cosmetic and drug manufacture, due to the extremely toxic nature

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service