Skip to Content
Merck
All Photos(2)

Key Documents

209074

Sigma-Aldrich

Ammonium hexachloroosmate(IV)

reagent grade

Synonym(s):

Ammonium osmium chloride, Osmium(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
€625.00

€625.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
€625.00

About This Item

Linear Formula:
(NH4)2OsCl6
CAS Number:
Molecular Weight:
439.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
reagent grade
form:
powder

€625.00


Check Cart for Availability

Request a Bulk Order

grade

reagent grade

Quality Level

form

powder

mp

170 °C (subl.) (lit.)

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[NH4+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2H3N.Os/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

SRBXXQDKBKTWOC-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Measurement of the dynamics of residual stresses in copper during heating: This study discusses the formation of PdOs bimetallic nanoalloy using ammonium hexachloroosmate(IV) and its application in understanding stress dynamics in metals during heating processes (ResearchGate).
  • Electrochemical DNA Hybridization Sensor Using Poly -co-Ethylamine Redox Polymer: This research discusses the preparation of Os(bpy)2Cl2 by mixing bipyridine with ammonium hexachloroosmate in ethylene glycol, showing the application of this compound in biosensor technology (Core.ac.uk).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chee Shan Lim et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(9), 1898-1905 (2015-04-25)
Renewable energy sources are highly sought after as a result of numerous worldwide problems concerning the environment and the shortage of energy. Currently, the focus in the field is on the development of catalysts that are able to provide water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service