Skip to Content
Merck
All Photos(2)

Key Documents

208183

Sigma-Aldrich

Cadmium sulfide

powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdS
CAS Number:
Molecular Weight:
144.48
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

synthesis grade

form

powder

mol wt

Mw 144.5 g/mol(lit.)

reaction suitability

reagent type: catalyst
core: cadmium

mp

−1750 °C (at a pressure of 100 bar) (lit.)

solubility

H2O: insoluble(lit.)

density

4.82 g/mL at 25 °C (lit.)

SMILES string

S=[Cd]

InChI

1S/Cd.S

InChI key

CJOBVZJTOIVNNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: hawleyite (structural type of sphalerite) and greenockite (structural type of wurtzite)

Application

Applications: pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries; phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

Packaging

Packaged in glass bottles

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janusz Szeremeta et al.
Nanoscale, 5(6), 2388-2393 (2013-02-12)
Wide wavelength range (550-1600 nm) measurements of nonlinear optical properties of ca. 5 nm CdS quantum dots (QDs) colloidal solution were performed using the Z-scan technique with femtosecond laser pulses at 1 kHz. Parameters characterizing nonlinear absorption and nonlinear refraction
Ou Chen et al.
Nature materials, 12(5), 445-451 (2013-02-05)
High particle uniformity, high photoluminescence quantum yields, narrow and symmetric emission spectral lineshapes and minimal single-dot emission intermittency (known as blinking) have been recognized as universal requirements for the successful use of colloidal quantum dots in nearly all optical applications.
Shancheng Yan et al.
Journal of nanoscience and nanotechnology, 13(6), 4242-4246 (2013-07-19)
A roll-on transfer strategy is developed to enable large-scale and uniform assembly of CdS nanowires on various rigid and flexible substrate materials. In this method, the CdS nanowires were synthesized by the hydrothermal method. The dispersed CdS NWs could be
Sandrine Ithurria et al.
Journal of the American Chemical Society, 134(45), 18585-18590 (2012-10-16)
Atomic layer deposition (ALD) is widely used for gas-phase deposition of high-quality dielectric, semiconducting, or metallic films on various substrates. In this contribution we propose the concept of colloidal ALD (c-ALD) for synthesis of colloidal nanostructures. During the c-ALD process
C Javaux et al.
Nature nanotechnology, 8(3), 206-212 (2013-02-12)
Applications of semiconductor nanocrystals such as biomarkers and light-emitting optoelectronic devices require that their fluorescence quantum yield be close to 100%. However, such quantum yields have not been obtained yet, in part, because non-radiative Auger recombination in charged nanocrystals could

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service