Skip to Content
Merck
All Photos(1)

Key Documents

20644

Sigma-Aldrich

O-tert-Butyl-L-threonine

≥98.0% (T)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€79.70

€79.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€79.70

About This Item

Empirical Formula (Hill Notation):
C8H17NO3
CAS Number:
Molecular Weight:
175.23
Beilstein:
2206127
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€79.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (T)

optical activity

[α]20/D −43±2°, c = 1% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O

InChI

1S/C8H17NO3/c1-5(6(9)7(10)11)12-8(2,3)4/h5-6H,9H2,1-4H3,(H,10,11)/t5-,6+/m1/s1

InChI key

NMJINEMBBQVPGY-RITPCOANSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sajjad Ahmad et al.
Drug design, development and therapy, 15, 1299-1313 (2021-04-02)
Organocatalytic asymmetric Michael addition is a strong approach for C-C bond formation. The objective of the study is to design molecules by exploiting the efficiency of Michael Adducts. We proceeded with the synthesis of Michael adducts by tailoring the substitution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service