Skip to Content
Merck
All Photos(1)

Key Documents

203661

Sigma-Aldrich

Lutetium (III) oxide

99.99% trace metals basis

Synonym(s):

Cassiopeium oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€380.00

€380.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€380.00

About This Item

Linear Formula:
Lu2O3
CAS Number:
Molecular Weight:
397.93
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

€380.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lutetium

density

9.42 g/mL at 25 °C (lit.)

SMILES string

O=[Lu]O[Lu]=O

InChI

1S/2Lu.3O

InChI key

MPARYNQUYZOBJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

A new AB2O4 phase, LuPd2O4, with potential uses in superconducting material as well as being of goechemical importance, was formed at elevated pressure and temperature from Lu2O3 and PdO in the presence of potassium chlorate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen, B-H.; Walker, D.
Chemistry of Materials, 9, 1700-1700 (1997)

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service