Skip to Content
Merck
All Photos(1)

Key Documents

20260

Sigma-Aldrich

(2R,6R)-2-tert-Butyl-6-methyl-1,3-dioxan-4-one

≥98.5% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O3
CAS Number:
Molecular Weight:
172.22
Beilstein:
4307891
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D −56±2°, c = 1% in chloroform

mp

82-84 °C (lit.)
82-84 °C

storage temp.

−20°C

SMILES string

C[C@@H]1CC(=O)O[C@@H](O1)C(C)(C)C

InChI

1S/C9H16O3/c1-6-5-7(10)12-8(11-6)9(2,3)4/h6,8H,5H2,1-4H3/t6-,8-/m1/s1

InChI key

SFWUTHYTQUXJTB-HTRCEHHLSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Derivative of (R)-β-hydroxy-butyric acid which can be α-alkylated in high anti-selectivity; 5-alkylidenation and transformation of products

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service