Skip to Content
Merck
All Photos(1)

Key Documents

201383

Sigma-Aldrich

Chicago Sky Blue 6B

Dye content 50 %

Synonym(s):

Direct Blue 1, Pontamine sky blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H24N6Na4O16S4
CAS Number:
Molecular Weight:
992.80
Colour Index Number:
24410
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 50%

λmax

618 nm

SMILES string

[Na+].[Na+].[Na+].[Na+].COc1cc(ccc1\N=N\c2ccc3c(cc(c(N)c3c2O)S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(\N=N\c5ccc6c(cc(c(N)c6c5O)S([O-])(=O)=O)S([O-])(=O)=O)c(OC)c4

InChI

1S/C34H28N6O16S4.4Na/c1-55-23-11-15(3-7-19(23)37-39-21-9-5-17-25(57(43,44)45)13-27(59(49,50)51)31(35)29(17)33(21)41)16-4-8-20(24(12-16)56-2)38-40-22-10-6-18-26(58(46,47)48)14-28(60(52,53)54)32(36)30(18)34(22)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4/b39-37+,40-38+;;;;

InChI key

BPHHNXJPFPEJOF-GPTZEZBUSA-J

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeinhom M El-Bahy et al.
Journal of hazardous materials, 166(1), 138-143 (2008-12-23)
Lanthanide ions (La(3+), Nd(3+), Sm(3+), Eu(3+), Gd(3+), and Yb(3+))/doped TiO2 nanoparticles were successfully synthesized by sol-gel method. Their photocatalytic activities were evaluated using Direct Blue dye (DB53) as a decomposition objective. The structural features of TiO2 and lanthanide ions/TiO2 were
New contrast stain for the rapid diagnosis of dermatophytic and candidal dermatomycoses.
Christopher Seng-Hong Lim et al.
Archives of dermatology, 144(9), 1228-1229 (2008-09-17)
Yukiko Urayama et al.
Autonomic neuroscience : basic & clinical, 152(1-2), 35-40 (2009-09-29)
Like pain sensation, vomiting is induced by C afferent fibers of the vagus nerve. Therefore, it can be assumed to be suppressed by the descending inhibitory system (DIS) of pain. In this study we examined this possibility in immobilized decerebrated
G Kútvölgyi et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 81(4-6), 109-117 (2006-11-30)
A simple trypan blue-neutral red-Giemsa staining procedure for simultaneous evaluation of acrosome, sperm head, and tail membrane integrity and morphology has been used to evaluate equine spermatozoa. Some special characteristics and problems have arisen in evaluating stallion semen. One problem
Isabel Mesquita et al.
Journal of hazardous materials, 237-238, 30-37 (2012-09-12)
In this work, oxidation with a Fenton-like process of a dye solution was carried out in a packed-bed reactor. Activated carbon Norit RX 3 Extra was impregnated with ferrous sulfate and used as catalyst (7 wt.% of iron). The effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service