Skip to Content
Merck
All Photos(1)

Key Documents

200956

Sigma-Aldrich

Hexafluorophosphoric acid solution

~55 wt. % in H2O

Synonym(s):

Hydrogen hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€43.50
500 G
€121.00

€43.50


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
€43.50
500 G
€121.00

About This Item

Linear Formula:
HPF6
CAS Number:
Molecular Weight:
145.97
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22
concentration:
~55 wt. % in H2O

€43.50


Estimated to ship on27 March 2025


Request a Bulk Order

form

liquid

Quality Level

concentration

~55 wt. % in H2O

density

1.651 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

F.FP(F)(F)(F)F

InChI

1S/F5P.FH/c1-6(2,3,4)5;/h;1H

InChI key

OQWOLAMXNGPSQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexafluorophosphoric acid (HPF6) is a strong acid that is commonly used as a catalyst in organic synthesis. It is also used in the polymerization and preparation of its salts.

Application

Hexafluorophosphoric acid solution is used as a catalyst for the conversion of polyethyleneimine (PEI) microgel into the corresponding polymeric ionic liquids (PIL) forms via protonation and anion exchange reaction. Polymeric ionic liquids are used as metal-free catalysts for hydrogen production via sodium borohydride methanolysis.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Product Class 12: Telluropyrylium and Benzotelluropyrylium Salts
Murphy PJ et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations null
The Use of Hexafluorophosphoric Acid in the Schiemann Reaction
Rutherford KG, et al.
The Journal of Organic Chemistry, 26(12), 5149-5152 (1961)

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service