Skip to Content
Merck
All Photos(1)

Documents

19990

Sigma-Aldrich

tert-Butyl hydroperoxide solution

~80% in di-tert-butyl peroxide/water 3:2

Synonym(s):

1,1-Dimethylethyl hydroperoxide, 2-Hydroperoxy-2-methylpropane, TBHP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COOH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098280
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

reaction suitability

reagent type: oxidant

concentration

~80% in di-tert-butyl peroxide/water 3:2

refractive index

n20/D 1.397

density

0.90 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OO

InChI

1S/C4H10O2/c1-4(2,3)6-5/h5H,1-3H3

InChI key

CIHOLLKRGTVIJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Org. Perox. C - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

109.4 °F

Flash Point(C)

43 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong Tang et al.
Ultrasonics sonochemistry, 20(5), 1168-1175 (2013-03-30)
This work investigated the ultrasonic assisted oxidative desulfurization of bunker-C oil with TBHP/MoO3 system. The operational parameters for the desulfurization procedure such as ultrasonic irradiation time, ultrasonic wave amplitude, catalyst initial concentration and oxidation agent initial concentration were studied. The
Yury Minko et al.
Nature protocols, 8(4), 749-754 (2013-03-23)
This protocol describes a new approach for the preparation of stereodefined trisubstituted chiral enolate species, avoiding conventional asymmetric enolization of carbonyl compounds. This protocol was developed as a single-flask synthetic sequence and therefore does not require isolation or purification of
Qicai Xue et al.
Chemical communications (Cambridge, England), 49(35), 3700-3702 (2013-03-29)
A new synthetic approach toward direct C-N bond formation through sp(3) C-H activation has been developed under metal-free conditions. Both primary and secondary benzylic C-H substrates could react smoothly with various amines to give only mono-amination products with good to
Younghwa Kim et al.
Food chemistry, 137(1-4), 136-141 (2012-12-04)
Oligomeric and polymeric procyanidins have been reported to possess different antioxidant capacities. However, the intracellular antioxidant mechanisms of oligomeric and polymeric procyanidins are still poorly understood. In this study, we evaluated the cytoprotective effects of the oligomeric procyanidin fraction (OPF)
Michael K Coggins et al.
Journal of the American Chemical Society, 135(11), 4260-4272 (2013-02-26)
Manganese-peroxos are proposed as key intermediates in a number of important biochemical and synthetic transformations. Our understanding of the structural, spectroscopic, and reactivity properties of these metastable species is limited, however, and correlations between these properties have yet to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service