Skip to Content
Merck
All Photos(1)

Key Documents

199338

Sigma-Aldrich

1,4-Diamino-2-butanone dihydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2COCH2NH2 · 2HCl
CAS Number:
Molecular Weight:
175.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

221 °C (dec.) (lit.)

functional group

amine
ketone

SMILES string

Cl[H].Cl[H].NCCC(=O)CN

InChI

1S/C4H10N2O.2ClH/c5-2-1-4(7)3-6;;/h1-3,5-6H2;2*1H

InChI key

HDRKYQXFDHZZOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Diamino-2-butanone dihydrochloride was used as a media supplement for the growth of B.pertussis and B. bronchiseptica.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukihiro Ueno et al.
Biological & pharmaceutical bulletin, 27(6), 890-892 (2004-06-10)
Candida albicans generally grows in hyphae form in RPMI1640 medium. However, addition of 1,4-diamino-2-butanone (DAB), a competitive inhibitor of ornithine decarboxylase, decreased the amount of polyamines in C. albicans, and induced the proliferation of the C. albicans yeast form. The
A G Tkachenko et al.
Mikrobiologiia, 70(4), 487-494 (2001-09-18)
The level of expression of oxyR, the gene that protects Escherichia coli against oxidative stress, was enhanced by physiological concentrations of the biogenic amine putrescine. This level was directly proportional to the degree of negative DNA supercoiling. 1,4-Diamino-2-butanone (DAB), a
A G Tkachenko et al.
Biochemistry. Biokhimiia, 72(1), 109-116 (2007-02-21)
Mechanisms of antioxidant effect of polyamines were studied in dependence on the strength of superoxide stress. Under conditions of weak stress, polyamines from Escherichia coli cultures were shown to function mainly as a scavenger of free superoxide radicals, whereas under
M Martinez-Pacheco et al.
Archives of microbiology, 151(1), 10-14 (1989-01-01)
Diamino butanone (DAB), a competitive inhibitor of ornithine decarboxylase (ODC) a key enzyme in polyamine biosynthesis, inhibited the yeast to hyphae transition in Mucor rouxii, induced by transfer from anaerobiosis to aerobiosis, but not the opposite phenomenon. Addition of DAB
D Guzmán-de-Peña et al.
Fungal genetics and biology : FG & B, 21(2), 198-205 (1997-04-01)
Regulation of aflatoxin biosynthesis during differentiation of Aspergillus parasiticus was analyzed by using a drug that inhibits the development of the fungus and mutants affected in sporulation. Diaminobutanone, a competitive inhibitor of ornithine decarboxylase, repressed spore germination. If added after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service