Skip to Content
Merck
All Photos(1)

Key Documents

199206

Sigma-Aldrich

Methyl 4-methoxyphenylacetate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€74.50

€74.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€74.50

About This Item

Linear Formula:
CH3OC6H4CH2CO2CH3
CAS Number:
Molecular Weight:
180.20
Beilstein:
2209665
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€74.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.516 (lit.)

bp

158 °C/19 mmHg (lit.)

density

1.135 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)Cc1ccc(OC)cc1

InChI

1S/C10H12O3/c1-12-9-5-3-8(4-6-9)7-10(11)13-2/h3-6H,7H2,1-2H3

InChI key

ZQYLDVNTWDEAJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 4-methoxyphenylacetate was used in the synthesis of 7,8-dihydroxy-2-(4′-hydroxybenzyl)-4H-1-benzopyran-4-one[1].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giulio Rastelli et al.
Bioorganic & medicinal chemistry, 10(5), 1427-1436 (2002-03-12)
The relative binding affinities to human aldose reductase (ALR2) of three new 7-hydroxy-2-benzyl-4H-1-benzopyran-4-one inhibitors were predicted by free energy perturbation (FEP) simulations. Molecular substitutions were specifically designed to investigate the role of hydrogen bonding at the active site of ALR2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service