Skip to Content
Merck
All Photos(1)

Key Documents

196924

Sigma-Aldrich

2-Amino-5-ethyl-1,3,4-thiadiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3S
CAS Number:
Molecular Weight:
129.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

200-203 °C (lit.)

SMILES string

CCc1nnc(N)s1

InChI

1S/C4H7N3S/c1-2-3-6-7-4(5)8-3/h2H2,1H3,(H2,5,7)

InChI key

QXTRPGAMVIONMK-UHFFFAOYSA-N

General description

Influence of 2-amino-5-ethylthio-1,3,4-thiadiazole on inhibition of copper corrosion in an aerated 0.50M HCl solution has been investigated using gravimetric and electrochemical techniques[1].

Application

2-Amino-5-ethyl-1,3,4-thiadiazole was used in the synthesis of:
  • 3-(5-ethyl-1,3,4-thiadiazol-2-yldiazenyl)-1-methyl-2-phenyl-1H-indole[2]
  • hetarylazoindole dyes[3]
  • N-(5-ethyl-[1,3,4]-thiadiazole-2-yl) toluenesulfonamide ligand[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis, spectroscopic properties and crystal structure of novel, bis-hetarylazo disperse dyes.
Seferoglu Z, et al.
Dyes and Pigments, 77(3), 614-625 (3008)
3-(5-Ethyl-1, 3, 4-thiadiazol-2-yldiazenyl)-1-methyl-2-phenyl-1H-indole.
Seferoglu Z, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(2), o568-o570 (2007)
Adriana Hangan et al.
Acta crystallographica. Section B, Structural science, 66(Pt 6), 615-621 (2010-11-26)
The crystal structure solution of the title compound is determined from microcrystalline powder using a multi-technique approach that combines X-ray powder diffraction (XRPD) data analysis based on direct-space methods with information from (13)C solid-state NMR (SSNMR), and molecular modelling using
Effects of 2-amino-5-ethylthio-1, 3, 4-thiadiazole on copper corrosion as a corrosion inhibitor in aerated acidic pickling solutions.
Sherif EM and Park S-M.
Electrochimica Acta, 51(28), 6556-6562 (2006)
Damien Cressier et al.
Bioorganic & medicinal chemistry, 17(14), 5275-5284 (2009-06-09)
In this work, we report the synthesis and characterization of new compounds derived from benzothiazoles and thiadiazoles. We observed that structural modifications on these skeletons affected the antioxidant activity. Thiol and aminothiol compounds derived from thiadiazoles and benzothiazoles showed an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service