Skip to Content
Merck
All Photos(1)

Key Documents

196150

Sigma-Aldrich

2-Iodothiophene

98%

Synonym(s):

α-Iodothiophene, 2-Thienyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3IS
CAS Number:
Molecular Weight:
210.04
Beilstein:
104666
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.651 (lit.)

bp

73 °C/15 mmHg (lit.)

mp

−40 °C (lit.)

density

1.902 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Ic1cccs1

InChI

1S/C4H3IS/c5-4-2-1-3-6-4/h1-3H

InChI key

ROIMNSWDOJCBFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mechanism of the fluoride-free, palladium-catalyzed cross-coupling reaction of 2-iodothiophene with potassium (E)-heptenyldimethylsilanolate has been investigated. 2-Iodothiophene participates in microwave-enhanced, rapid homogeneous-phase version of the Sonogashira reaction.

Application

2-Iodothiophene was used in the synthesis of (E)-and (Z)-2,3-difluoro-3-aryl substituted acrylic esters. It was also used as monomer in the preparation of thin conductive films via plasma polymerization of 2-iodothiophene.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott E Denmark et al.
Journal of the American Chemical Society, 126(15), 4876-4882 (2004-04-15)
The mechanism of the fluoride-free, palladium-catalyzed cross-coupling reaction of potassium (E)-heptenyldimethylsilanolate, K(+)(E)-1(-), with 2-iodothiophene has been investigated through kinetic analysis. The order of each component was determined by plotting the initial rates of the reaction against concentration. These data provided
M Erdélyi et al.
The Journal of organic chemistry, 66(12), 4165-4169 (2001-06-09)
A microwave-enhanced, rapid and efficient homogeneous-phase version of the Sonogashira reaction is presented. It has been applied to the coupling of aryl iodides, bromides, triflates, and aryl chloride, as well as pyridine and thiophene derivatives with trimethylsilylacetylene. Excellent yields (80-95%)
Thin conductive coatings formed by plasma polymerization of 2-iodothiophene.
Kruse A, et al.
Surface and Coatings Technology, 59(1), 359-364 (1993)
Barbara Marchetti et al.
The Journal of chemical physics, 142(22), 224303-224303 (2015-06-15)
Velocity map imaging methods, with a new and improved ion optics design, have been used to explore the near ultraviolet photodissociation dynamics of gas phase 2-bromo- and 2-iodothiophene molecules. In both cases, the ground (X) and spin-orbit excited (X*) (where
Yi Wang et al.
The Journal of organic chemistry, 70(26), 10743-10746 (2005-12-17)
[reaction: see text] The (Z)-2,3-difluoro-3-stannylacrylic ester is readily prepared from (Z)-1,2-difluorovinyltriethylsilane via stereospecific stannyl/silyl exchange with KF/(Bu3Sn)2O or Bu3SnCl in DMF at 70 degrees C. The corresponding (E)-2,3-difluoro-3-stannylacrylate is prepared by stereospecific carbonylation of (E)-1,2-difluorovinyl iodide followed by low temperature/in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service