Skip to Content
Merck
All Photos(1)

Key Documents

19597

Sigma-Aldrich

4-(3-Butyl-1-imidazolio)-1-butanesulfonic acid triflate

≥97.0%

Synonym(s):

1-Butyl-3-(4-sulfobutyl)imidazolium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21F3N2O6S2
CAS Number:
Molecular Weight:
410.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0%

impurities

≤3.0% water
≤50 mg/kg halogens (as chloride)

anion traces

bromide (Br-): 50 mg/kg
chloride (Cl-): 50 mg/kg
fluoride (F-): 50 mg/kg
nitrate (NO3-): 50 mg/kg
sulfate (SO42-): 2000 mg/kg

SMILES string

[O-]S(=O)(=O)C(F)(F)F.CCCCn1cc[n+](CCCCS(O)(=O)=O)c1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vivian R Feig et al.
Nature communications, 9(1), 2740-2740 (2018-07-18)
Conductive and stretchable materials that match the elastic moduli of biological tissue (0.5-500 kPa) are desired for enhanced interfacial and mechanical stability. Compared with inorganic and dry polymeric conductors, hydrogels made with conducting polymers are promising soft electrode materials due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service