Skip to Content
Merck
All Photos(1)

Key Documents

194387

Sigma-Aldrich

2-Chlorobenzophenone

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€57.60

€57.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€57.60

About This Item

Linear Formula:
ClC6H4COC6H5
CAS Number:
Molecular Weight:
216.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.60


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

bp

330 °C (lit.)

mp

44-47 °C (lit.)

functional group

chloro
ketone
phenyl

SMILES string

Clc1ccccc1C(=O)c2ccccc2

InChI

1S/C13H9ClO/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9H

InChI key

VMHYWKBKHMYRNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermodynamics of formation of inclusion complex between 2-chlorobenzophenone and cyclomaltoheptaose (β-cyclodextrin) has been investigated by UV-vis spectroscopy and reversed-phase liquid chromatography[1]. 2-Chlorobenzophenone undergoes reduction in the presence of LiAlH4 and (R)-(-)-2-(2-iso-indolinyl)butan-1-ol to afford the corresponding benzhydrols[2].

Application

2-Chlorobenzophenone was used in the synthesis of 1-(2-chlorophenyl)isoquinolin-3-yl trifluoromethanesulfonate[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of N-methyl-N-(1-methylpropyl)-1-(2-chlorophenyl) isoquinoline-3-[11 C] carboxamide ([11 C-carbonyl] PK11195) and some analogues using [11 C] carbon monoxide and 1-(2-chlorophenyl) isoquinolin-3-yl triflate.
Rahman O, et al.
Journal of the Chemical Society. Perkin Transactions 1, 23, 2699-2703 (2002)
Asymmetric reductions of ketones using lithium aluminium hydride modified with N, N-dialkyl derivatives of (R)-(-)-2-aminobutan-1-ol.
Brown E, et al.
Tetrahedron Asymmetry, 2(5), 339-342 (1991)
Matias I Sancho et al.
Carbohydrate research, 346(13), 1978-1984 (2011-06-07)
A thermodynamic study of the inclusion process between 2-chlorobenzophenone (2ClBP) and cyclomaltoheptaose (β-cyclodextrin, β-CD) was performed using UV-vis spectroscopy, reversed-phase liquid chromatography (RP-HPLC), and molecular modeling (PM6). Spectrophotometric measurements in aqueous solutions were performed at different temperatures. The stoichiometry of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service