192988
3-Chloropropyl thiolacetate
technical grade, ≥90%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
technical grade
Assay
≥90%
form
liquid
refractive index
n20/D 1.495 (lit.)
bp
83-84 °C/10 mmHg (lit.)
density
1.159 g/mL at 25 °C (lit.)
SMILES string
CC(=O)SCCCCl
InChI
1S/C5H9ClOS/c1-5(7)8-4-2-3-6/h2-4H2,1H3
InChI key
IOLUCUQNAZTKMN-UHFFFAOYSA-N
General description
Mechanism of formation of trimethylene oxide from 3-chloropropyl thiolacetate has been investigated.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
172.4 °F - closed cup
Flash Point(C)
78 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mechanism of Trimethylene Oxide Formation from 3-Chloropropyl Acetate.
Journal of the American Chemical Society, 79(16), 4431-4435 (1957)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service