Skip to Content
Merck
All Photos(1)

Documents

192732

Sigma-Aldrich

Diethylaluminum chloride solution

25 wt. % in toluene

Synonym(s):

Chlorodiethylaluminum, Diethylaluminium chloride, Diethylaluminum chloride (6CI), Diethylaluminum monochloride, Diethylchloroaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2AlCl
CAS Number:
Molecular Weight:
120.56
Beilstein:
4123259
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

25 wt. % in toluene

density

0.887 g/mL at 25 °C

SMILES string

CC[Al](Cl)CC

InChI

1S/2C2H5.Al.ClH/c2*1-2;;/h2*1H2,2H3;;1H/q;;+1;/p-1

InChI key

YNLAOSYQHBDIKW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service