Skip to Content
Merck
All Photos(1)

Documents

191493

Sigma-Aldrich

4-Chloro-1,8-naphthalic anhydride

technical grade

Synonym(s):

6-Chloronaphtho[1,8,8a-c,d]pyran-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5ClO3
CAS Number:
Molecular Weight:
232.62
MDL number:

grade

technical grade

mp

202-206 °C (dec.)

SMILES string

Clc1ccc2C(=O)OC(=O)c3cccc1c23

InChI

1S/C12H5ClO3/c13-9-5-4-8-10-6(9)2-1-3-7(10)11(14)16-12(8)15/h1-5H

InChI key

UJEUBSWHCGDJQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service