Skip to Content
Merck
All Photos(1)

Documents

189553

Sigma-Aldrich

Poly(vinyl chloride) carboxylated

average Mw ~220,000 by GPC, powder

Synonym(s):

Poly(vinyl chloride-co-acrylic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2 · C2H3Cl)x
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

carboxylated

form

powder

mol wt

average Mw ~220,000 by GPC

extent of labeling

1.8% carboxyl

inherent viscosity

1.4 dL/g(lit.)

transition temp

Tg 84 °C

solubility

acetone/carbon disulfide, MEK, THF: soluble

SMILES string

ClC=C

InChI

1S/C3H4O2.C2H3Cl/c1-2-3(4)5;1-2-3/h2H,1H2,(H,4,5);2H,1H2

InChI key

SQNNHEYXAJPPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Warning

May contain vinyl chloride, an OSHA carcinogen. See 29 CFR part 1910

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IIIrd International Symposium on Neural Transplantation. Cambridge, 6-11 August, 1989. Proceedings.
Progress in brain research, 82, 1-734 (1990-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service