Skip to Content
Merck
All Photos(2)

Key Documents

188271

Sigma-Aldrich

4-Chloro-2-nitrobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€110.00

€110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€110.00

About This Item

Linear Formula:
ClC6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
187.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€110.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

90-92 °C (lit.)

functional group

chloro
hydroxyl
nitro

SMILES string

OCc1ccc(Cl)cc1[N+]([O-])=O

InChI

1S/C7H6ClNO3/c8-6-2-1-5(4-10)7(3-6)9(11)12/h1-3,10H,4H2

InChI key

OAHGNOHGHOTZQU-UHFFFAOYSA-N

General description

4-Chloro-2-nitrobenzyl alcohol undergoes oxidation in the presence of pyridinium chlorochromate in dichloromethane to yield 4-chloro-2-nitrobenzaldehyde[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyaza heterocycles. Part 2. Nucleophilic substitution of halogens in halogenoquinoxalino [2, 3-c] cinnolines.
Ahmad A, et al.
Journal of the Chemical Society. Perkin Transactions 1, 19, 2751-2758 (1994)
Brian Dobosh et al.
STAR protocols, 2(4), 100892-100892 (2021-11-09)
Neutrophils are difficult to study, particularly in tissues, due to their short half-life and propensity for activation. We describe an organotypic airway model that uses patient airway fluid to enable the transmigration of blood neutrophils to acquire an airway-like phenotype

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service