Skip to Content
Merck
All Photos(2)

Documents

185493

Sigma-Aldrich

N-Benzylaniline

≥99%

Synonym(s):

N-Phenylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHC6H5
CAS Number:
Molecular Weight:
183.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

solid

bp

306-307 °C (lit.)

mp

35-38 °C (lit.)

solubility

alcohol: soluble
chloroform: soluble
diethyl ether: soluble
water: insoluble

density

1.061 g/mL at 25 °C (lit.)

SMILES string

C(Nc1ccccc1)c2ccccc2

InChI

1S/C13H13N/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13/h1-10,14H,11H2

InChI key

GTWJETSWSUWSEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The electropolymerisation of N-benzylaniline at transparent Indium Tin Oxide glass electrodes has been investigated by UV-visible spectroelectrochemistry. N-Benzylaniline on electrochemical oxidation in aqueous sulfuric acid solution produces an adherent conducting polymer film at the platinum electrode.

Application

N-Benzylaniline was used in the separation of tervalent gallium, indium and thallium by solvent extraction method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A UV-visible spectroelectrochemical study of the electropolymerisation of N-benzylaniline.
Malinauskas A and Holze R.
Journal of Solid State Electrochemistry, 3(7-8), 429-436 (1999)
M M Khosla et al.
Talanta, 21(6), 411-415 (1974-06-01)
A simple and rapid method is proposed for the separation of tervalent gallium, indium and thallium by solvent extraction with N-benzylaniline in chloroform from different concentrations of hydrochloric acid. Thallium and gallium are extracted from 1M and 7.0-7.5M hydrochloric acid
M Ulgen et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(8), 735-748 (1994-08-01)
1. The in vitro hepatic microsomal metabolism of certain substituted N-benzylanilines was studied in the male hamster to establish the mechanism(s) and process(es) involved in the formation of the corresponding amides. 2. N-Benzyl-2,4,6-trihalogeno, N-benzyl-4-cyano- and N-benzyl-4-nitroanilines were only metabolized by
Synthesis and structure-activity relationships of a class of sodium iodide symporter function inhibitors.
Fanny Waltz et al.
ChemMedChem, 6(10), 1775-1777 (2011-07-14)
Yung-Hung Chang et al.
Dalton transactions (Cambridge, England : 2003), (5)(5), 861-867 (2009-01-22)
Both saturated and unsaturated N-benzyl substituted heterocyclic carbene (NHC) iridum(i) complexes were synthesized. The unsaturated carbene complex [(un-NHC-Bn)Ir(CO)(2)Cl] in the cis form was prepared via the carbene transfer from the corresponding silver complex to [Ir(COD)(2)Cl](2) followed by ligand substitution with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service