Skip to Content
Merck
All Photos(1)

Documents

185175

Sigma-Aldrich

1-Benzyl-3-pyrrolidinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
Beilstein:
1526217
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.539 (lit.)

bp

77 °C/0.01 mmHg (lit.)

density

1.091 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CCN(C1)Cc2ccccc2

InChI

1S/C11H13NO/c13-11-6-7-12(9-11)8-10-4-2-1-3-5-10/h1-5H,6-9H2

InChI key

DHGMDHQNUNRMIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The equilibrium constant for the ketoreductase-catalyzed reduction reaction of 1-benzyl-3-pyrrolidinone has been measured in n-hexane. 1-Benzyl-3-pyrrolidinone on enzymatic asymmetric reduction yields enantiopure 1-benzyl-3-hydroxypyrrolidine, well known intermediate for various drugs.

Application

1-Benzyl-3-pyrrolidinone was used as starting reagent in the synthesis of vinyl triflate. It was used to prepare chiral, alkenyl sulfoximines leading to highly functionalized diazabicycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis of 3-arylpyrroles by tandem Suzuki-dehydrogenation reaction.
Lee C-W and Chung YJ.
Tetrahedron Letters, 41(18), 3423-3425 (2000)
Synthesis, 2224-2224 (2006)
Practical synthetic process for enantiopure 1-benzyl-3-hydroxypyrrolidine.
Morimoto M and Sakai K.
Tetrahedron Asymmetry, 19(12), 1465-1469 (2008)
A thermodynamic study of ketoreductase-catalyzed reactions 5. Reduction of substituted ketones in n-hexane.
Tewari YB, et al.
The Journal of Chemical Thermodynamics, 40(4), 661-670 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service