Skip to Content
Merck
All Photos(1)

Key Documents

185140

Sigma-Aldrich

3,3′-Dichloropivalic acid

≥97%

Synonym(s):

2,2-Bis(chloromethyl)propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€132.00

€132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€132.00

About This Item

Linear Formula:
CH3C(CH2Cl)2CO2H
CAS Number:
Molecular Weight:
171.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€132.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

form

solid

mp

64-66 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

CC(CCl)(CCl)C(O)=O

InChI

1S/C5H8Cl2O2/c1-5(2-6,3-7)4(8)9/h2-3H2,1H3,(H,8,9)

InChI key

DDSPBKFTRPWDLI-UHFFFAOYSA-N

Related Categories

General description

3,3′-Dichloropivalic acid reacts with ethane-1,2-diamine to yield isomeric tetra-amine derivatives, tetra amino carboxylic acid and carboxamidotriamino alcohol[1].

Application

3,3′-Dichloropivalic acid was used in the synthesis of 3,3′-dichloropivaloyl chloride[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul V Bernhardt et al.
Inorganic chemistry, 43(5), 1681-1688 (2004-03-03)
Reaction between ethane-1,2-diamine and 3,3'-dichloropivalic acid results in different, isomeric tetra-amine derivatives, one a tetraamino carboxylic acid and the other a carboxamidotriamino alcohol, depending upon reaction conditions. Intended conversion of the Cu(II) complex of the former to a cyclam-like macrocycle
A novel rearrangement reaction conversion of 3-(chloromethyl) azetidin-2-ones to azetidine-3-carboxylic acid esters.
Bartholomew D and Stocks MJ.
Tetrahedron Letters, 32(36), 4795-4798 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service