Skip to Content
Merck
All Photos(1)

Key Documents

184675

Sigma-Aldrich

3-Methylstyrene

99%, contains 0.1% 3,5-di-tert-butylcatechol as inhibitor

Synonym(s):

3-Vinyltoluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€134.00

€134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€134.00

About This Item

Linear Formula:
CH3C6H4CH=CH2
CAS Number:
Molecular Weight:
118.18
Beilstein:
1304618
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€134.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

contains

0.1% 3,5-di-tert-butylcatechol as inhibitor

refractive index

n20/D 1.541 (lit.)

bp

170-171 °C (lit.)

mp

−82-−81 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C=C)c1

InChI

1S/C9H10/c1-3-9-6-4-5-8(2)7-9/h3-7H,1H2,2H3

InChI key

JZHGRUMIRATHIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yasuhito Suzuki et al.
Forensic science international, 278, 228-239 (2017-08-02)
Various autopsy findings are used for the diagnosis of burning death, including inhalation injury, soot in the respiratory tract, vital reaction, blood carboxyhemoglobin (COHb), and blood cyanide. However, not all findings are necessarily complete, and autopsy diagnosis can be difficult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service